Learn More
Lovibond™ Ammonia Reagent Set Powder Packs™
Premeasured unit dose reagent for water quality testing
Brand: Lovibond™ 535500
Code : NEW
Additional Details : Weight : 0.30000kg Transport : UN number : 2680 Chem class : 8 Pack group : II
Includes: Ammonia Cyanurate Reagent (10mL sample, 100 pack), Ammonia Salicylate Reagent (10mL sample, 100 pack)
Description
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
10mL ammonia cyanurate reagent (100 pack), 10mL ammonia salicylate reagent (100 pack) | |
0 to 0.8mg/L | |
Ammonia Nitrogen Reagent Set |
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers | |
100 | |
Salicylate |
Safety and Handling
- vario ammonia nitrogen set
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : lithium hydroxide monohydrate, sodium dichloroisocyanurate,dihydrate, sodium salicylate, sodium nit