missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Total Phosphate Reagent Set, for Tube Testing

Premeasured unit dose reagent for water quality testing

Brand:  Lovibond™ 535210


Product Code. 10364153

  • £106.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Acid Reagent Vial (50ct), Phosphate 3 Reagent (10mL sample, 50 pack), Potassium Persulfate Reagent (10mL sample, 50 pack), Sodium hydroxide (1.54 N, 100mL), Sodium hydroxide (1.00 N, 100mL), Deionized water (100mL)
This item is not returnable. View return policy

Description

Description

  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Specifications

Specifications

Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
50 ct. acid reagent vial, 10mL phosphate 3 reagent (50 pack), 10mL potassium persulfate reagent (50 pack), 1.54N 100mL sodium hydroxide, 100mL deionized water
0.02 to 1.1mg/L P/0.06 to 3.5mg/L PO4
Total Phosphate Reagent Set
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers, reactor
50
Acid digestion/Ascorbic Acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • vario total phosphate reagent set

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 1 A
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H331-Toxic if inhaled.
  • H290-May be corrosive to metals.
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P234-Keep only in original container.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P390-Absorb spillage to prevent material damage.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, disodium peroxodisulphate, potassium persulphate, sodium hydroxide, dipotassium disu

SDS
Documents

Documents

Certificates
Special Offers

Special Offers