Learn More
Lovibond™ Tablet Reagent: Hardness BW
Guarantees exact reagent dosing thereby allowing for more precise titrations
Brand: Lovibond™ 515171BT
Code : NEW
Additional Details : Weight : 0.30000kg Transport : UN number : 3259 Chem class : 8 Pack group : II
Description
- Convenient blister packaging allows user to push tablet through foil directly into sample
- Each tablet is hermetically sealed in an individual aluminum foil pocket with shelf life of five years
- Use with LR Total Hardness MINIKIT
Specifications
Routine water analysis | |
LR total hardness minikit | |
Hardness BW Tablet |
Safety and Handling
- PASTILLE durete LR BW
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 2-amino-2-methylpropane-1,3-diol, tri-sodium phosphate dodecahydrate