missing translation for 'onlineSavingsMsg'
Learn More

GE Analytical Instruments™ Low Capacity 150mL Oxidizer Reagent Cartridge

For use with the Sievers M-Series, 900 and 5310 C Series.

Brand:  GE Analytical Instruments™ APF 90150-02


Product Code. 15384067

  • £506.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Replace after 3 months of use
  • For use with TOC instrumentation
  • Designed and built to exact specifications for GE Analytical Instruments’ analyzers and sensors
  • Raw material purity managed through rigorous testing and control

Specifications

Specifications

Certificate of Analysis
150 mL
Oxidation
Sievers M-Series
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffered Ammonium Persulfate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic

SDS
Documents

Documents

Certificates
Special Offers

Special Offers