missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drierite™ Drying Tubes, Cole-Parmer™
Click to view available options
Quantity:
4 Pk.
Packaging:
4 Pack
Description
Tubes dry to a dew point of –100°F with a flow rate up to 300mL/min. Drying tubes measure 8 in. L x 3/4 in. OD and come filled with 30 grams of indicating Drierite.
- Inlet/outlet ports accept 1/4 in. to 3/8 in. ID tubing
- 20 micron Porex™ filter is in each end
Specifications
Specifications
Material | Calcium Sulfate with Cobalt Chloride |
Quantity | 4 Pk. |
Packaging | 4 Pack |
Pore Volume | 20 |
Safety and Handling


Product Identifier
- INDICATING DRIERITE
- Danger
- Respiratory sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H411-Toxic to aquatic life with long lasting effects.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
- MIXTURE LIST-Contains : cobalt dichloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Drierite™ Drying Tubes, Cole-Parmer™ >
Spot an opportunity for improvement?Share a Content Correction