Learn More
3M™ Particulate Respirator, Series C100
Provide comfortable, effective respiratory protection against dust and mist. The 3M C100 Series Particulate Respirator is lightweight and comfortable.
£14.60 - £22.65
Specifications
Material Polypropylene
View More Specs
Products 4
Product Code Brand Filter Type Product Type Price Quantity & Availability  
Product Code Brand Filter Type Product Type Price Quantity & Availability  
13587000
View Documents
3M™
7000088840
FFP1 NR D Unvalved Particulate Respirator
£19.90
Pack of 20
In Stock
to see stock.
Edge
Add to basket
13597000
View Documents
3M™
7000088842
FFP2 NR D Unvalved Particulate Respirator This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13507010
View Documents
3M™
7000088841
FFP1 NR D Valved Particulate Respirator
£14.60
Pack of 10
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
13517010
View Documents
3M™
7000088843
FFP2 NR D Valved Particulate Respirator This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Provides cost-effective, lightweight, comfortable, and effective protection against dust and mist.
  • The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes
  • Exhalation valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
Specifications
Specifications
Polypropylene