Learn More
Kartell™ Low Form Beaker with Blue Graduations
Accurately measures the volume of a liquid. Kartell™ Low Form Beaker with Blue Graduations uses horizontal marked lines to assist in measuring out various volumes of a liquid.
£16.60 - £127.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10654432
|
Kartell™
0172000 |
25 mL |
£71.30
Pack of 20 |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10555811
|
Kartell™
0172200 |
100 mL |
£57.45
Pack of 12 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
10041071
|
Kartell™
0172300 |
250 mL |
£127.00
Pack of 16 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
10431765
|
Kartell™
0172400 |
500 mL |
£16.60
Each |
In Stock
Log in to see stock. |
|||||
10063734
|
Kartell™
0172500 |
1000 mL |
£21.30
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
10691742
|
Kartell™
0172600 |
2000 mL |
£85.20
Pack of 4 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
10601752
|
Kartell™
0172700 |
3000 mL |
£64.65
Pack of 2 |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10070941
|
Kartell™
0172800 |
5000 mL |
£119.00
Pack of 2 |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
Description
Accurately measures the volume of a liquid. Kartell™ Low Form Beaker with Blue Graduations uses horizontal marked lines to assist in measuring out various volumes of a liquid.
- Good chemical resistance
- Permanent blue graduations moulded into beaker
- Can withstand temperatures up to 170°C for short periods
- Suitable for applications with food
Specifications
PMP | |
Autoclavable | |
Yes |