Learn More
Thermo Scientific Chemicals Low Form Inconel Crucible
Brand:  Thermo Scientific Chemicals 035886.KT
 View more versions of this product
Product Code. 15423585
  • £64.82 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Low Form
15 mL
23mm
25mm
Inconel
33mm
Inconel Crucible