missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Low Profile Tubes and Domed Caps

Low profile strip tubes for PCR and qPCR applications.

Brand:  Thermo Scientific™ AB-0847

 View more versions of this product

Product Code. 11514804

This item is not returnable. View return policy

Description

Description

Ideal for applications requiring low reaction volumes, Thermo Scientific™ Low Profile Strip Tubes are compatible with 0.2mL cycler blocks.

The low profile design of the Thermo Scientific Low Profile Strip Tubes reduces the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Thermo Scientific low profile products are especially recommended for use with reaction volumes below 20 μL.

The low profile design of the Thermo Scientific strip tubes without caps reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL.

Highlights

  • Suitable for 0.2 mL thermal cycler blocks
  • 150 μL maximum tube capacity when capped
  • Low profile to reduce dead space and increase PCR efficiency
  • Labelled A-H end tabs (8-strip) and 1-12 end tabs (12-strip)
Applications
  • PCR
  • qPCR/Real-Time PCR

Hazardous: No
Storage Condition: Ambient

PCR, qPCR and Real-Time PCR.

Certifications

Certified free from human DNA, RNases, and DNases.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified free from human DNA, RNases, and DNases
Natural
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers