missing translation for 'onlineSavingsMsg'
Learn More

Low Temperature Viscosity Standard, Paragon Scientific™

Product Code. p-7161122
Click to view available options
Viscosity:
151216.0 mPa.s (-40°C)
172195 mPa.s (-13°C)
5989.0 mPa.s (-40°C)
757.0 mPa.s (-40°C)
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15886461

Brand: Paragon Scientific J10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paragon ISO 17025 / ISO 17034 certified low temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.

  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
  • Specifications

    Chemical Name or Material J10
    Viscosity 757.0 mPa.s (-40°C)
    Certifications/Compliance ISO 17025, UKAS ISO 17034
    Quantity 500 mL
    compliance-icons compliance-icons
    Product Identifier
    • Viscosity Reference Standard Low Temperature Type
    Signal Word
    • Danger
    Hazard Category
    • LONG-TERM AQUATIC HAZARD Chronic 3
    • Aspiration toxicity Category 1
    • Skin corrosion/irritation Category 2
    • Specific target organ toxicity Category 3
    Hazard Statement
    • H304-May be fatal if swallowed and enters airways.
    • H315-Causes skin irritation.
    • H336-May cause drowsiness or dizziness.
    • H412-Harmful to aquatic life with long lasting effects.
    Precautionary Statement
    • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
    • P264-Wash skin thoroughly after handling.
    • P273-Avoid release to the environment.
    • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
    • P302+P352-IF ON SKIN: Wash with plenty of water/.
    • P331-Do NOT induce vomiting.
    Supplemental information
    • MIXTURE LIST-Contains : Odourless Kerosene

    Nominal values quoted

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.