Learn More
Miele Professional™ Lower Basket A 150
Lower basket A 150 for the optimum loading of injector modules or inserts
Brand:  Miele Professional™ 9862400
577.50 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 15382579
  • £1035.00 / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lower basket with two module connections
  • for holding up to 2 injector modules or inserts
  • Automatic closing of the connections when not in use
  • Clearances: width 494, length 440 mm
Specifications
Specifications
Stainless Steel
Stainless Steel
PG 8583/PG8593 laboratory washers
546 mm
529 mm
Basket
Lower basket for modules
Two docking pipes, self-sealing docking valves
154 mm