missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Loxistatin

Product Code. 15427099
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

A cell-permeable, irreversible inhibitor of cysteine proteases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88321-09-9
Molecular Formula C17H30N2O5
Synonym EST; E-64d

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A cell-permeable, irreversible inhibitor of cysteine proteases
Formula Weight 342.43
Solubility Soluble in DMSO, DMF or ethanol
Form Powder
InChI Key SRVFFFJZQVENJC-IHRRRGAJSA-N
SMILES CCOC(=O)[C@H]1O[C@@H]1C(=O)N[C@@H](CC(C)C)C(=O)NCCC(C)C
IUPAC Name ethyl (2S,3S)-3-{[(1S)-3-methyl-1-[(3-methylbutyl)carbamoyl]butyl]carbamoyl}oxirane-2-carboxylate
Molecular Weight (g/mol) 342.44
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'