missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek LP610NM Chromaglo Red Emitting Luciferase
For use with microplate readers and imagers
Brand: Agilent BioTek 7092209
This item is not returnable.
View return policy
Description
- Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
- Fluorescence filter for use with single and multimode microplate readers and imagers
Specifications
Microplate Readers and Imagers |