Learn More
LRR 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11628-1-AP-20UL
 View more versions of this product
Product Code. 16878833
  • £135.00 / 20µL
Explore more special offers
Description
Description
May negatively regulate the 4-1BB-mediated signaling cascades which result in the activation of NK-kappaB and JNK1. Probable substrate recognition subunit of an ECS (Elongin BC- CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications
Specifications
LRR 1
Polyclonal
Unconjugated
LRR1
4 1BBLRR, LRR 1, LRR repeat protein 1, LRR1, PPIL5
Rabbit
Antigen Affinity Chromatography
RUO
122769, 214345, 685860
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q80VQ1, Q96L50
Lrr1, Lrrc1
LRR 1 Fusion Protein Ag2159
20 μL
Primary
Mouse, Rat, Human