Learn More
LRRC14B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558227
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LRCIEFPVPK DCYPEGAAYP QDELAMSKFN QQKYDEIAEE LRAVLLRADR EDIQVSTPLF GSFDPDIQET SNELGAFLLQ AFKTAL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
The protein encoded by this gene is a leucine-rich repeat containing protein that is a member of the PRAME family.Specifications
LRRC14B | |
Polyclonal | |
Unconjugated | |
LRRC14B | |
AI595366; leucine rich repeat containing 14B; leucine-rich repeat-containing protein 14B; LRRC14B; RGD1564978 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
389257 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NHZ5 | |
LRRC14B | |
Recombinant protein corresponding to Human LRRC14B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only