missing translation for 'onlineSavingsMsg'
Learn More
LRRK2-IN-1, Tocris Bioscience™ Product Code.: 18025622

LRRK2-IN-1, Tocris Bioscience™

Product Code. 18025622
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18025622

Brand: Tocris Bioscience 4273/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective LRRK2 inhibitor

Potent and selective inhibitor of leucine-rich repeat kinase 2 (LRRK2). Inhibits both G2019S mutant and wild-type LRRK2 kinase activity (IC50 values are 6 and 13 nM respectively). Causes dephosphorylation, ubiquitination and degradation of LRRK2. Inhibits IFN-γ-induced monocyte maturation in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1234480-84-2
Molecular Formula C31H38N8O3
Molecular Weight (g/mol) 570.698
InChI Key IWMCPJZTADUIFX-UHFFFAOYSA-N
Synonym lrrk2-in-1, lrrk2in1, lrrk2-in1, 2-2-methoxy-4-4-4-methylpiperazin-1-yl piperidine-1-carbonyl phenyl amino-5,11-dimethyl-5h-benzo e pyrimido 5,4-b 1,4 diazepin-6 11h-one, 2-2-methoxy-4-4-4-methylpiperazin-1-yl piperidine-1-carbonyl phenyl-amino-5,11-dimethyl-5h-benzo e pyrimido 5,4-b 1,4 diazepin-6 11h-one, 2-2-methoxy-4-4-4-methylpiperazin-1-yl piperidine-1-carbonyl anilino-5,11-dimethylpyrimido 4,5-b 1,4 benzodiazepin-6-one, 2-2-methoxy-4-4-4-methylpiperazin-1-yl piperidin-1-yl carbonyl phenyl amino-5,11-dimethyl-5,11-dihydro-6h-pyrimido 4,5-b 1,4 benzodiazepin-6-one, 5,11-dihydro-2-2-methoxy-4-4-4-methyl-1-piperazinyl-1-piperidinyl carbonyl phenyl amino-5,11-dimethyl-6h-pyrimido 4,5-b 1,4 benzodiazepin-6-one, lrrk-2in1, lrrk2-tn-1
PubChem CID 46843906
ChEBI CHEBI:78413
IUPAC Name 2-[2-methoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]anilino]-5,11-dimethylpyrimido[4,5-b][1,4]benzodiazepin-6-one
SMILES CN1CCN(CC1)C2CCN(CC2)C(=O)C3=CC(=C(C=C3)NC4=NC=C5C(=N4)N(C6=CC=CC=C6C(=O)N5C)C)OC

Specifications

Quantity 10 mg
Formula Weight 570.09
Percent Purity >98%
Chemical Name or Material LRRK2-IN-1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
LRRK2-IN-1, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.