missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LST1 293T Cell Overexpression Lysate (Denatured), Abnova
Brand: Abnova H00007940-T01.100uL
This item is not returnable.
View return policy
Description
Western Blotting
Specifications
293T | |
LST1 293T Cell Transient Overexpression Lysate(Denatured) | |
13.09 | |
pCMV-LST1 full-length | |
100 μL | |
Human |
Western Blot | |
NM_007161 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-LST1 antibody (H00007940-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |