Learn More
Grant Instruments™ ecocool™ Refrigerated/Heated Circulating Bath
GREENER_CHOICE
Saves up to 80% against standard compressor units. Grant Instruments™ ecocool™ Refrigerated/Heated Circulating Baths are supplied assembled as ready to use kits, complete with accessory hosing, clips and connectors as standard.
Brand:  Grant Instruments™ ECOCOOL 100R
3706.68 GBP valid until 2024-12-31
Use promo code "21805" to get your promotional price.
 View more versions of this product
Product Code. 15499214
  • £4160.00 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Eco mode optimises the cooling control needed, enabling rapid cool down when required.
  • True energy saving against competitor units that only switch the compressor on or off.
  • Intuitive user interface
  • No-spill valved front drain.
  • Built in Britain to the highest specifications, 5 day dispatch, 4 year industry leading warranty.
  • Modern, sleek, attractive design
  • Thermostat and chiller work in harmony, neither will operate alone, eliminating any danger of overheating or freezing.
  • Single front switch for user convenience.
  • No side vents - locate to suit the user, not the unit.
Specifications
Specifications
5 L
-20°C to +100°C
ecocool™ 100
0.05°C
245 mm
640 mm
29 kg
50/60 Hz
250W at 20°C, 230W at 10°C, 185W at 0°C, 115W at -10°C, 40W at -10°C
4 digit LED
3
4 Years
154 mm
118 mm
120/230 V