missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Lubricating Grease Molykote G4500 25 g
Lubricating Grease Molykote G4500
Brand: Thermo Scientific™ 75003786
27.69 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Lubricating Grease Molykote G4500
- Lubricating Grease Molykote G4500 25g
- Grease for Trunnion and Thread of Rotors.

Specifications
Lubricating Grease | |
Trunnion and Thread of Rotors |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Lubricating Grease Molykote G4500 25 g > Trunnion and Thread of Rotors
Spot an opportunity for improvement?Share a Content Correction