Learn More
Invitrogen™ Luc-Screen™ Extended-Glow Luciferase Reporter Gene Assay System
Description
Includes
Two reaction buffers
- Homogeneous assay format allows detection of firefly luciferase in presence of normal culture media without removal of media and without an additional cell lysis step, providing easiest and most streamlined assay procedure possible
- Glow assay kinetics provides a window during which measurements may be performed, facilitating HTS applications where assay automation is used
- Wide dynamic range of firefly luciferase assay lets the user measure enzyme level accurately from femtogram to nanogram range
- Assay sensitivity is 100- to 1,000-fold better than with either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assay technologies
- Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
- Non-radioactive reporter gene assay kit eliminates radioisotopes
- Assay can be completed in about one hour, providing fast assay turnaround
Sensitive
Luciferase is an ideal reporter because of the high sensitivity of detection and the absence of endogenous luciferase activity in mammalian cells. The Luc-Screen assay conveniently couples in-well cell lysis in the presence of culture medium with a high sensitivity assay that exhibits extended-glow light emission kinetics. Light signal can be measured between 10 minutes and several hours after adding assay reagents, providing great flexibility in the time between reagent addition and measurement.
High Throughput
The Luc-Screen system is designed for maximum assay flexibility in a high throughput format and can be used in luminometers without automatic injectors. The Luc-Screen system is formulated to provide a convenient, easy-to-use luciferase assay that is optimized for use in high-throughput screening. The high sensitivity of the Luc-Screen assay is accompanied by a wide dynamic range.
Specifications
- Assay Type: Luciferase Assay
- Compatible Cells: Mammalian Cells
- Detection Method: Chemiluminescent
- For Use With Equipment: Luminometer (Microplate)
- Form: Liquid
- Format: 96-well plate
- Readout: End Point
- Sample Type (Specific): Cell Cultures
- Substrate: Luciferin
- Substrate Properties: Chemical Substrate
- Substrate Type: Luciferase Substrate
Specifications
Specifications
| Product Type | Luciferase Reporter Gene Assay System |
| Detection Method | Chemiluminescence |
| For Use With (Equipment) | Luminometer (Microplate) |
| Assay | Luciferase Assay |
| Content And Storage | This kit contains two reaction buffers. Store the product at 4°C. |
| Compatible Cells | Mammalian Cells |
| High-throughput Compatibility | High-throughput Compatible |
| Readout | End Point |
| Sample Type | Cell Cultures |
| Substrate | Luciferin |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.