missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luc-Screen™ Extended-Glow Luciferase Reporter Gene Assay System

Thermo Scientific Luc-Screen Extended-Glow Luciferase Reporter Gene Assay System with extended-glow light emission is designed for sensitive detection of firefly luciferase reporter enzyme in microplate cell cultures. A linear signal is obtained with the

£234.00 - £4820.00

Specifications

Product Type Luciferase Reporter Gene Assay System
Detection Method Chemiluminescence
For Use With (Equipment) Luminometer (Microplate)
Assay Luciferase Assay
Compatible Cells Mammalian Cells
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12624295
View Documents
Invitrogen™
T1033
1000 Assays
£700.00
Each
Please to purchase this item. Need a web account? Register with us today!
12634295
View Documents
Invitrogen™
T1035
200 Assays
£234.00
Each
Please to purchase this item. Need a web account? Register with us today!
12614295
View Documents
Invitrogen™
T1034
10,000 Assays
£4820.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Luciferase Reporter Gene Assay System
Luminometer (Microplate)
Mammalian Cells
End Point
Luciferin
Chemical Substrate
Luc-Screen™, NovaBright™
Liquid
Chemiluminescence
Luciferase Assay
High-throughput Compatible
Cell Cultures
Luciferase Substrate
Luciferase
Enzyme Assay
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Luc-Screen™ Extended-Glow Luciferase Reporter Gene Assay System

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.