missing translation for 'onlineSavingsMsg'
Learn More
Lucifer yellow CH, Dilithium salt, ≥99%, >95%, MP Biomedicals Product Code.: 11477532
  • ONLINE EXCLUSIVE PRICE
    ÂŁ304.00
    ÂŁ239.00 / 25mg
    Save ÂŁ65.00
    21% Off

Lucifer yellow CH, Dilithium salt, ≥99%, >95%, MP Biomedicals

Product Code. p-4495715 Shop All MP Biomedicals Products
25 mg, 25mg
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 11477532

Brand: MP Biomedicals 0215526725

  • ONLINE EXCLUSIVE PRICE
    ÂŁ304.00
    ÂŁ239.00 / 25mg
    Save ÂŁ65.00
    21% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67769-47-5
Molecular Formula C13H9Li2N5O9S2
Molecular Weight (g/mol) 457.241
InChI Key RPKCZJYDUKVMGF-UHFFFAOYSA-L
Synonym 6-amino-2[(Hyrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-Benz[de] Isoquinoline-5,8-disulfonic acid dilithium salt
PubChem CID 124220108
ChEBI CHEBI:52106
IUPAC Name dilithium;6-amino-2-(hydrazinecarbonylamino)-1,3-dioxobenzo[de]isoquinoline-5,8-disulfonate
SMILES [Li+].[Li+].C1=C(C=C2C3=C1C(=O)N(C(=O)C3=CC(=C2N)S(=O)(=O)[O-])NC(=O)NN)S(=O)(=O)[O-]

Specifications

Assay Percent Range >95%
Formula Weight 457.2
Percent Purity ≥99%
Color Orange to Yellow
Quantity 25 mg
Physical Form Powder
Chemical Name or Material Lucifer yellow CH, Dilithium salt, For HPLC analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lucifer yellow CH, Dilithium salt, ≥99%, >95%, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.