Learn More
Cole Parmer Luer Fittings
Cole Parmer Luer Fittings feature exceptional durability and are easy to disassemble for cleaning
Marca: Cole Parmer 41507-26
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Cole Parmer Luer Fittings feature exceptional durability and are easy to disassemble for cleaning
Ideal for harsh chemical environmentsEspecificaciones
Luer fittings, male luer lock x 1/16 in. hose barb | |
Autoclavable | |
5/8 in. | |
1 Ea. |
Male luer lock x 1/16 in. hose barb | |
Stainless Steel | |
15.748 mm | |
5/8 in. |