missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BD Combi™ Plug
Click to view available options
For Use With (Equipment):
Catheters, Infusion Lines, Luer Syringes
Unit Size:
Pack of 100
Description
- Luer-Lok cap with male/female protection
- To proteg/plug catheters and intravenous infusion lines
- With latex-free membrane for carrying out injections with a needle
- For medical use - Orange polypropylene
- Individually wrapped—100 per box
Specifications
Specifications
| Material | Polypropylene |
| Color | Orange |
| Description | Luer-Lok cap with male/female protection |
| For Use With (Application) | For medical use |
| For Use With (Equipment) | Catheters, Infusion Lines, Luer Syringes |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction