Learn More
Thermo Scientific™ Luer-Tip, Gas-Tight Manual Syringes for GC Systems
Get a gas-tight syringe for manual injections. This syringe features a Luer-Tip connection and is available in a variety of sizes.
£88.25 - £135.00
Product Code | Brand | Volume (Metric) | Needle Gauge | Length (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Length (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||
10371195
|
Thermo Scientific™
36502025 |
5 μL | 26 G | 80 mm | N/A | ||||
12886026
|
Thermo Scientific™
365C0231 |
5 μL | 23s G | 57 mm | TriPlus RSH, TriPlus 100LS Autosamplers |
£90.10
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15274593
|
Thermo Scientific™
365K7817 |
1000 μL | Gas Chromatography Systems |
£95.48
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13416856
|
Thermo Scientific™
365D0261 |
10 μL | 23s G | 85 mm | TriPlus RSH Autosampler |
£91.76
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13446856
|
Thermo Scientific™
365D0281 |
10 μL | 26s G | 85 mm | TriPlus RSH Autosampler |
£90.10
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15214603
|
Thermo Scientific™
365N7820 |
10 mL | Gas Chromatography Systems |
£135.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13426856
|
Thermo Scientific™
365D0301 |
10 μL | 23s G | 85 mm | TriPlus RSH Autosampler |
£112.56
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12806036
|
Thermo Scientific™
365D0311 |
10 μL | 23s G | 57 mm | TriPlus RSH Autosampler |
£88.25
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Get a gas-tight syringe for manual injections in your GC system. This syringe features a Luer-Tip connection and is available in a variety of sizes. Made of quality materials, they are designed for easy, accurate, precise and reproducible injections.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.