missing translation for 'onlineSavingsMsg'
Learn More

Luff-Schoorl Reagent, Honeywell Fluka™

Volumetric, 25 g/L

Brand:  Honeywell Fluka 34400-1L

Code : NEW

Additional Details : CAS Number : 497-19-8 Weight : 1.15700kg Transport : UN number : 3082 Chem class : 9 Pack group : III

 View more versions of this product

Product Code. 15624170

  • £56.86 / 1 L
Estimated Shipment: 24-10-2024
to see stock.

Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Specifications

Luff-Schoorl Reagent
497-19-8,7732-18-5,7758-98-7
UN3082
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers