Learn More
Luff-Schoorl Reagent, Honeywell Fluka™
Volumetric, 25 g/L
Brand: Honeywell Fluka 34400-5L-VP
Code : NEW
Additional Details : Weight : 5.00000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Luff-Schoorl Reagent | |
497-19-8,7732-18-5,7758-98-7 | |
UN3082 | |
5 L |