Learn More
Thermo Scientific™ Luminaris Color Probe qPCR Master Mix, high ROX
Ready-to-use qPCR master mixes with color and premixed UDG optimized for probe detection using standard cycling protocols.
£220.00 - £3200.00
Specifications
Description | Color Probe High ROX |
---|---|
Detection Method | Primer-probe |
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13273159
|
Thermo Scientific™
K0331 |
250 x 20 μL Reactions |
£220.00
250 reactions |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13283159
|
Thermo Scientific™
K0332 |
500 x 20 μL Reactions |
£423.00
500 reactions |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13203169
|
Thermo Scientific™
K0333 |
1250 x 20 μL Reactions |
£1010.00
1250 reactions |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13213169
|
Thermo Scientific™
K0334 |
5000 x 20 μL Reactions |
£3200.00
5000 reactions |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific™ Luminaris Color Probe qPCR Master Mixes are ready-to-use qPCR master mixes with color and premixed UDG optimized for probe detection using standard cycling protocols.
Thermo Scientific Luminaris Color Probe qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR. The master mixes include Thermo Scientific Hot Start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimized PCR buffer. Only the template, primers and probe need to be added.The Hot Start Taq DNA polymerase, in combination with an optimized buffer, ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control. The Luminaris Color Probe qPCR Master Mixes are supplemented with an inert blue dye and a separate Yellow Sample Buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.
The use of Luminaris Color Probe qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color Probe qPCR Master Mixes with ROX at different concentrations as passive reference dye are available for most qPCR platforms.
Specificity no primer dimers
Sensitivitity detects low copy number targets
Wide linear range accurate quantification across 9 orders of magnitude
Reproducibility and convenience ready-to-use 2X master mix
- Blue Master Mix and Yellow Sample Buffer for easy pipetting
- UDG in the master mix to prevent carry-over contamination
- Excellent qPCR performance
- Versions with premixed ROX for different real-time platforms
- Standard cycling protocols
Includes:
- Luminaris Color Probe qPCR Master Mixes include Hot Start Taq Polymerase, UDG and dNTPs (also dUTP) in an optimized PCR buffer with a blue dye
- 40X Yellow Sample Buffer with a yellow dye
- Nuclease-free water
Recommended for:
Gene expression analysis; siRNA validation; Genotyping; Pathogen detection
Specifications
Color Probe High ROX | |
Tube | |
qPCR | |
Standard | |
Luminaris | |
Gene Expression, Genotyping, siRNA Validation | |
DNA, dsDNA |
Primer-probe | |
High | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System | |
qPCR Master Mix | |
2X (Luminaris Color Probe High ROX qPCR Master Mix), 40X (Yellow Sample Buffer) |
For Research Use Only. Not for use in diagnostic procedures.