Learn More
Thermo Scientific™ Luminaris Color Probe qPCR Master Mix
Optimize qPCR with probe detection using standard cycling protocols with these ready-to-use master mixes with premixed UDG, available with or without blue color.
£225.00 - £3295.00
Specifications
Description | Color Probe |
---|---|
Detection Method | Primer-probe |
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
13273169
|
Thermo Scientific™
K0353 |
1,250 Reactions |
£1044.00
1250 reactions |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13293169
|
Thermo Scientific™
K0351 |
250 Reactions |
£225.00
250 reactions |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13283169
|
Thermo Scientific™
K0354 |
5,000 Reactions |
£3295.00
5000 reactions |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13263169
|
Thermo Scientific™
K0352 |
500 reactions |
£427.00
500 reactions |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Luminaris Color Probe and Luminaris Probe qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR. The master mixes include Thermo Scientific Hot Start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimized PCR buffer. Only the template, primers and probe need to be added.
The Hot Start Taq DNA polymerase, in combination with an optimized buffer, ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control. The Luminaris Color Probe qPCR Master Mixes are supplemented with an inert blue dye and a separate Yellow Sample Buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.
The use of Luminaris Color Probe and Luminaris Probe qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color Probe and Luminaris Probe qPCR Master Mixes with ROX at different concentrations as passive reference dye are available for most qPCR platforms.
Highlights
- Blue Master Mix and Yellow Sample Buffer for easy pipetting
- UDG in the master mix to prevent carry-over contamination
- Excellent qPCR performance
- Specificity — Hot Start Taq DNA Polymerase and the optimized buffer reduce primer dimers
- Sensitivitity — single copy detection
- Wide linear range — accurate quantification across 9 orders of magnitude
- Reproducibility and convenience — ready-to-use 2X master mix
- Equal performance of blue and colorless master mixes
- Versions with premixed ROX for different real-time platforms
- Standard cycling protocols
Specifications
Color Probe | |
Tube | |
qPCR | |
Standard | |
Luminaris | |
Gene Expression | |
DNA, dsDNA |
Primer-probe | |
High | |
Taq DNA Polymerase | |
BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon | |
qPCR Master Mix | |
2X (Luminaris Color Probe qPCR Master Mix), 40X (Yellow Sample Buffer) |
For Research Use Only. Not for use in diagnostic procedures.