missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Luminex 200™ Calibration Kit
Click to view available options
Quantity:
25 uses
Unit Size:
25 reactions
Description
A lot-specific CD is also included for importing target values. The kit is intended to be used with the Automated Maintenance Plate (AMP). Stripwells for the AMP are included with each kit. Each kit will provide 25 calibrations.
Specifications
Specifications
| Type | Calibration Kit |
| Product Line | Luminex |
| Quantity | 25 uses |
| Content And Storage | • 28 Strips wells • L x 100/200 calibration kit CD • Luminex classification calibrator microspheres • MagPlex classification calibrator microspheres • Luminex reporter calibrator microspheres Store at 2°C to 8°C. |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction