Learn More
Thermo Scientific™ Accessories for Luminoskan™ Ascent Microplate Luminometer
Run a variety of luminometric research applications with this compact, robust instrument certified as DLR-Ready.
£9660.00
Code produit | Marque | Prix | Quantité et disponibilité | ||||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Prix | Quantité et disponibilité | ||||||
12413458
|
Thermo Scientific™
2805621 |
1st Dispenser Kit for Luminoskan |
£9660.00
Each |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Run a variety of luminometric research applications with the Thermo Scientific™ Luminoskan™ Ascent Microplate Luminometer, a compact and robust instrument offering versatile plate formats, fast reading speeds and up to three dispensers. This instrument has passed the DLReady™ validation at Promega Corporation™ and is certified as DLReady. The Luminoskan Ascent Microplate Luminometer is validated for Thermo Scientific™ Pierce™ Luciferase Assays.
Fast reading speed:
- Reads a 96-well plate in just 15 seconds
- Essential for kinetic applications, such as enzyme kinetics and phagocytosis assays
Onboard dispensers for exact follow-up of kinetic reactions:
- Equipped with up to three reagent dispensers for fast kinetic assays, such as ATP assays
- Supports simultaneous dispensing and reading, enabling monitoring of fast kinetic measurements from the very start of the reaction
- Offers very low dead volume and backflush capability for conservation of expensive reagents
High sensitivity:
- DLReady certified for excellent sensitivity
- Fiberless direct illumination optics ensures maximal light collection
- High sensitivity, a wide dynamic range and low crosstalk for accurate and precise results in luminometric assays
Specially designed for automation:
- Reads up to 384-well plates for increased assay throughput
- Easily integrates with automated systems
Includes:
PC Software
Recommended for:
- Reporter gene assays
- Immunoassays with luminescent substrates
- Cell proliferation and cytotoxicity assays
- Intracellular Ca2+ assays
- ATP assays
- Phagocytosis assays
- Reactive oxygen assays
- Microbiological assays
- Enzyme assays
- BRET assays
- ADMEtox
Prestations de service
1 year
Spécification
Luminoskan | |
PC Software |