Learn More
American Educational Products Lung/Breath Volume Kits
Investigate pulmonary fitness.
Brand:  American Educational Products 6109
 View more versions of this product
Product Code. 13142191
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Lung Volume Kit allows you to measure lung volume with reusable components
  • Breath Volume Kit measures tidal and respiratory minute volume
Specifications
Specifications
Breath Volume Kit
8 to 15
Durable plastic bag, mouthpiece and complete instruction guide
3 to 9
To measure lung volume with reusable components
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
American Educational Products Lung/Breath Volume Kits > Breath volume kit