Learn More
American Educational Products Lung/Breath Volume Kits
Investigate pulmonary fitness.
Brand: American Educational Products 6050
This item is not returnable.
View return policy
Description
- Lung Volume Kit allows you to measure lung volume with reusable components
- Breath Volume Kit measures tidal and respiratory minute volume
Specifications
Lung Volume Kit | |
3 to 9 | |
To measure lung volume with reusable components | |
Durable plastic bag calibrated to 6l, mouthpiece and mouthpiece holder |