missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lung Tissue Slides (Adenocarcinoma)- Paraffin
Shop All Bio Techne Products
Click to view available options
Species:
Human
Unit Size:
Pack of 5
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
| Cross Reactivity | Human |
| Type | Lung Tissue Slides (Adenocarcinoma-Moderately Differentiated Papillary) |
| For Use With (Application) | Immunohistochemistry-Paraffin |
| Regulatory Status | This product is for research use only and is not approved for use in humans or in clinical diagnosis. |
| Species | Human |
| Tissue Condition | Adenocarcinoma |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction