Learn More
Novus Biologicals™ Lung Tissue Slides (Normal)- Paraffin
Applications: Immunohistochemistry-Paraffin
Brand: Novus Biologicals™ NBP2-30182
Additional Details : Weight : 0.00970kg
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.Specifications
Human | |
Immunohistochemistry-Paraffin | |
Human |
Lung Tissue Slides (Normal) | |
This product is for research use only and is not approved for use in humans or in clinical diagnosis. | |
Normal |