missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPperCHIP™ Lupus Microarray
Lupus Microarray with 5062 antigen specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.
Brand: PEPperCHIP™ SPC.043.001
This item is not returnable.
View return policy
Description
- Microarray Content: 28 selected Lupus autoantigens and additional 789 linear Lupus epitopes of the Immune Epitope Database including posttranslational modifications like citrulline and acetyllysine
- Database Epitope Lengths: 3 to 50 aa
- Peptide Lengths: 3 to 15 aa
- Number of Peptides: 5062 printed in duplicate and HA, polio and c-Myc control peptides
Specifications
PEGMA/PMMA | |
1 Slide | |
1 mm | |
Transparent | |
Individual barcode for each microarray | |
High-density Peptide Microarray | |
Non-sterile |
1 array copy per microarray | |
Combinatorial peptide synthesis by amino acid laser printing | |
ISO 9001:2015 | |
RUO | |
Microscopic Glass | |
Antibody validation, epitope mapping, serum screening, biomarker discovery | |
75.4 x 25 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction