missing translation for 'onlineSavingsMsg'
Learn More

Luteolin, Tocris Bioscience™

Product Code. 18064062
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Anti-inflammatory, antioxidant and free radical scavenger

Anti-inflammatory, antioxidant and free radical scavenger. Inhibits LPS-induced TNF-α, IL-6 and inducible nitric oxide production and blocks NF-κB and AP-1 activation. Also inhibits TNF-α -induced COX-2 expression. Antiproliferative and chemopreventative; inhibits proliferation of Lewis lung carcinoma cells in vivo. Inhibits influenza virus replication in vitro by preventing viral entry. Molecular docking studies also indicate binding to SARS-CoV-2 main protease (Mpro or 3CLpro).

Chemical Identifiers

CAS 491-70-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.239
InChI Key IQPNAANSBPBGFQ-UHFFFAOYSA-N
Synonym luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide
PubChem CID 5280445
ChEBI CHEBI:15864
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one
SMILES C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O

Specifications

Quantity 50 mg
Formula Weight 286.24
Percent Purity >98%
Chemical Name or Material Luteolin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'