missing translation for 'onlineSavingsMsg'
Learn More

Lutetium Standard For ICP, 1000μg/mL, SPEX CertiPrep™

Catalog Number 10541183
Click to view available options
Quantity:
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Lu1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS 7439-94-3, 7697-37-2, 7732-18-5
Packaging Triple-leached LDPE bottle
Quantity 500 mL
CAS Max % 2.0, 0.1, 97.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula Lu
MDL Number MFCD00011098
InChI Key OHSVLFRHMCKCQY-UHFFFAOYSA-N
SMILES [Lu]
IUPAC Name lutetium
Molecular Weight (g/mol) 174.97
PubChem CID 23929
ChEBI CHEBI:33382
Grade Assurance
Concentration or Composition (by Analyte or Components) 1000mg/L
Analyte or Component Names Lu
Solvent or Matrix 2% HNO3
Chemical Name or Material Lutetium Standard, For AA and ICP
Show More Show Less
compliance-icons
Product Identifier
  • 1000 ug/mL Lutetium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'