missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Product Code. 11326328
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11326328

Brand: Thermo Scientific Alfa Aesar 011260.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15230-79-2 | H3LuN3O9 | 364.00 g/mol

Lutetium(III) chloride hexahydrate is used as a catalyst and laboratory reagent. It is also used in ceramics, optical glasses, electrical components and photo-optical material. It is one of the components of scintillating composition. It serves as a precursor for the preparation of other lutetium complexes. It finds application as one of the starting materials for aluminate phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15230-79-2
Molecular Formula H3LuN3O9
Molecular Weight (g/mol) 364.00
MDL Number MFCD00149775
InChI Key MESQZVHAWZZUAY-UHFFFAOYSA-N
IUPAC Name lutetium(3+) tris(nitric acid)
SMILES [Lu+3].O[N+]([O-])=O.O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 5 g
Merck Index 14,5615
Solubility Information Soluble in water.
Formula Weight 389.42 (281.33 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Lutetium(III) chloride hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.