missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific™

Product Code. 11364118
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-81-1
MDL Number MFCD00016191

Specifications

Melting Point 1182°C
Physical Form Pieces
Quantity 1 g
UN Number UN3288
Formula Weight 231.97
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 8.3 g/mL
Assay (REO)
Chemical Name or Material Lutetium(III) fluoride, Anhydrous
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'