missing translation for 'onlineSavingsMsg'
Learn More
Lutetium(III) fluoride, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11362578

Lutetium(III) fluoride, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11362578
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11362578

Brand: Thermo Scientific Alfa Aesar 014620.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-81-1
MDL Number MFCD00016191

Specifications

Melting Point 1182°C
Physical Form Lump
Quantity 5 g
UN Number UN3288
Formula Weight 231.97
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 8.3 g/mL
Assay (REO)
Chemical Name or Material Lutetium(III) fluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW23K054 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number014620.06
Lot NumberY10J035 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number014620.06,014620.03
Lot NumberR18J019 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number014620.06,014620.03
Lot NumberF20H18 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014620.06
Lot NumberP06F004 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014620.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lutetium(III) fluoride, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.