missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) fluoride, REacton™, 99.99% (REO), Thermo Scientific™

£47.52 - £179.00

Chemical Identifiers

CAS 13760-81-1
MDL Number MFCD00016191
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346328
SDS View Documents
Thermo Scientific Alfa Aesar
011261.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11356328
SDS View Documents
Thermo Scientific Alfa Aesar
011261.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13760-81-1
MFCD00016191

Specifications

1182°C
Lump
UN3288
99.99%
Odorless
8.3 g/mL
Lutetium(III) fluoride
13760-81-1
MFCD00016191
231.97
Reacton™
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 237-355-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lutetium(III) fluoride, REacton™, 99.99% (REO), Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.