missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11302518
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Lutetium(III) nitrate hydrate is used in ceramics, glass, phosphors and in lasers. It finds application as a catalyst in petroleum cracking and to prepare laser crystal. Further, it is used in hydrogenation, alkylation and polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100641-16-5
Molecular Formula LuN3O9
Molecular Weight (g/mol) 360.98
MDL Number MFCD00150421
InChI Key APRNQTOXCXOSHO-UHFFFAOYSA-N
Synonym lutetium iii nitrate hydrate, lutetium nitrate hydrate, acmc-1buji, lu.3no3.h2o, lutetium nitrate-water 1/3/1, lutetium 3+ hydrate trinitrate
PubChem CID 16212852
SMILES [Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 1 g
Linear Formula Lu(NO3)3·xH2O
UN Number UN1477
Solubility Information Soluble in water. Slightly soluble in strong mineral acids.
IUPAC Name lutetium(3+);trinitrate;hydrate
Formula Weight 360.98 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lutetium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.