Learn More
Lutetium(III) oxide, 99.99%, (trace metal basis), -325 mesh
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 12032-20-1 |
Color | White |
Physical Form | Powder |
Quantity | 1 g |
Assay Percent Range | 99.94% min |
Molecular Formula | Lu2O3 |
Linear Formula | Lu2O3 |
MDL Number | MFCD00011100 |
Merck Index | 14, 5615 |
Synonym | lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+ |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Safety and Handling
- Lutetium (III) oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.