missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) oxide, REacton™, 99.9% (REO)

Product Code. p-4408130
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11356318

Brand: Thermo Scientific Alfa Aesar 011255.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12032-20-1 | Lu2O3 | 397.93 g/mol

Lutetium(III) oxide serves as a catalyst in a variety of reactions such as polymerization, hydrogenation and alkylation reactions. It is also used in ceramics and glass. In the production of laser crystals, lutetium(III) oxide finds application as an important starting material. Its luminescent properties, a field of interest for researchers and may find potential application in biological fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-20-1
Molecular Formula Lu2O3
Molecular Weight (g/mol) 397.93
MDL Number MFCD00011100
InChI Key UGBIHFMRUDAMBY-UHFFFAOYSA-N
Synonym lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
IUPAC Name dilutetium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Lu+3].[Lu+3]

Specifications

Melting Point 2487°C
Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 397.94
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 9.42
Chemical Name or Material Lutetium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.