missing translation for 'onlineSavingsMsg'
Learn More
Lutetium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11391779

Lutetium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11391779
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11391779

Brand: Thermo Scientific Alfa Aesar 012410.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-94-3
Molecular Formula Lu
Molecular Weight (g/mol) 174.97
MDL Number MFCD00011098
InChI Key OHSVLFRHMCKCQY-UHFFFAOYSA-N
Synonym lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
PubChem CID 23929
ChEBI CHEBI:33382
IUPAC Name lutetium
SMILES [Lu]

Specifications

Color Silver
UN Number UN3178
Quantity 5 g
Percent Purity 99.9%
Odor Odorless
Form Pieces
Assay (REO)
Chemical Name or Material Lutetium pieces, Distilled dendritic

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ23K024 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number012410.03,012410.06
Lot NumberY05K020 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number012410.03,012410.06
Lot NumberZ02K026 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog Number012410.03,012410.06
Lot NumberX23K018 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number012410.03,012410.06
Lot NumberT25K021 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number012410.03,012410.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lutetium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.