missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Luxol Fast Blue MBSN, pure
£53.31 - £81.81
Chemical Identifiers
CAS | 1328-51-4 |
---|---|
Molecular Formula | C32H14CuN8Na2O6S2 |
Molecular Weight (g/mol) | 780.16 |
MDL Number | MFCD00071424 |
InChI Key | DKBXPLYSDKSFEQ-UHFFFAOYSA-L |
Synonym | Solvent Blue 38 |
PubChem CID | 92030797 |
SMILES | C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10348250
|
Thermo Scientific Chemicals
212170250 |
25 g | Glass bottle |
£53.31
25g |
In Stock
Log in to see stock. |
|||||
10591374
|
Thermo Scientific Chemicals
212171000 |
100 g | Glass bottle |
£81.81
100g |
In Stock
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1328-51-4 | |
780.16 | |
DKBXPLYSDKSFEQ-UHFFFAOYSA-L | |
92030797 |
C32H14CuN8Na2O6S2 | |
MFCD00071424 | |
Solvent Blue 38 | |
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2] |
Specifications
1328-51-4 | |
MFCD00071424 | |
Solubility in water: insoluble. Other solubilities: soluble in acohols like methanol, ethanol, propano, ls; very soluble in gylcols, ethanolamine, choline, . insoluble in hydrocarbons. | |
Authentic | |
780.16 | |
778.15 | |
Complies | |
Powder |
C32H14CuN8Na2O6S2 | |
Solvent Blue 38 | |
DKBXPLYSDKSFEQ-UHFFFAOYSA-L | |
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2] | |
92030797 | |
Pure | |
Blue | |
Luxol™ Fast Blue MBSN, Pure |
Safety and Handling
Product Identifier
- Luxol]TS Fast Blue MBSN
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Luxol Fast Blue MBSN, pure