missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LUZP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528172
This item is not returnable.
View return policy
Description
Recommended positive controls: PC-3, U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Export adapter involved in mRNA nuclear export in cancer cells. Binds and enhances the RNA-binding activity of the nuclear RNA export factor NXF1. Can restore mRNA export function in cells compromised by loss of mRNA export adapters.Specifications
LUZP4 | |
Polyclonal | |
Unconjugated | |
LUZP4 | |
Cancer/testis antigen 28; CT28; CT-28; CT-8; HOM-TES-85; HOM-TES-85 tumor antigen; leucine zipper protein 4; LUZP4; Tumor antigen HOM-TES-85 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51213 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9P127 | |
LUZP4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 264 of Human LUZP4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |