missing translation for 'onlineSavingsMsg'
Learn More

LY 367385, Tocris Bioscience™

Product Code. 30000712
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective mGlu1a antagonist

A selective mGlu1a receptor antagonist, with an IC50 value of 8.8 μM for blockade of quisqualate-induced phosphoinositide hydrolysis vs. > 100 μM for mGlu5a, and negligible action on group II and III receptors.

Chemical Identifiers

CAS 198419-91-9
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
InChI Key SGIKDIUCJAUSRD-QMMMGPOBSA-N
Synonym unii-d8uw47h17b, ly-367385 hydrochloride, chembl94631, 4-s-amino carboxy methyl-3-methylbenzoic acid, s-+-a-amino-4-carboxy-2-methylbenzeneacetic acid, 4-1s-1-amino-2-hydroxy-2-oxoethyl-3-methylbenzoic acid, tocris-1237, d04jqr, l-367385 analogs, lilly, metabotropic mglur1 agonists, lilly
PubChem CID 5311261
IUPAC Name 4-[(S)-amino(carboxy)methyl]-3-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)C(=O)O)C(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 209.2
Percent Purity >99%
Chemical Name or Material LY 367385
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'