missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6G Monoclonal Antibody (1A8-Ly6g), Brilliant Ultra Violet™ 395, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£160.00 - £319.00
Specifications
| Antigen | Ly-6G |
|---|---|
| Clone | 1A8-Ly6g |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17816871
|
Invitrogen™
363-9668-80 |
25 μg |
£160.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17826871
![]() |
Invitrogen™
363-9668-82 |
100 μg |
£319.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ly6G (Lymphocyte antigen 6 complex locus G6D) is a component of the myeloid differentiation antigen Gr-1, together with Ly6C. The Ly6G protein has a molecular weight of approximately 21-25 kDa and is expressed by myeloid derived cells in the bone marrow. Ly6G is a good marker for detection of peripheral neutrophils, monocytes and granulocytes. Expression of Gr-1 in bone marrow correlates with granulocyte differentiation and maturation. The physiological role of Ly6G remains unclear but studies have shown its involvement in anti-tumor response, and studies with neutrophils suggest that Ly6G ligation may have an inhibitory effect on local immune responses.Specifications
| Ly-6G | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Gr1; Gr-1; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6G | |
| Ly6g | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Ly6g |
| 1A8-Ly6g | |
| Flow Cytometry | |
| Brilliant Ultraviolet 395 | |
| Rat | |
| Mouse | |
| P35461 | |
| 546644 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title